Assay Details
| Gene Symbol: GPCPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019593.3 | NP_062539.1 | 2 - 3 | 266 | 119 | |
| XM_005260761.1 | XP_005260818.1 | 2 - 3 | 266 | 119 | ||
| GenBank mRNA | AB037855.1 | - | 2 - 3 | 257 | 119 | |
| AK308111.1 | - | 2 - 3 | 264 | 119 | ||
| BC027588.2 | - | 2 - 3 | 265 | 4827346 | 119 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB037855.1, BC027588.2, NM_019593.3, XM_005260761.1 transcripts.
Set Membership: |
|

GPCPD1
Genomic Map