Assay Details
| Gene Symbol: MFSD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294342.1 | NP_001281271.1 | 2 - 3 | 546 | 70 | |
| XM_005272297.1 | XP_005272354.1 | 2 - 3 | 639 | 70 | ||
| XM_006713919.1 | XP_006713982.1 | 2 - 3 | 639 | 70 | ||
| XM_011513572.1 | XP_011511874.1 | 2 - 3 | 726 | 70 | ||
| GenBank mRNA | AK025922.1 | - | 2 - 3 | 378 | 70 | |
More Information
Set Membership: |
|

MFSD7
Genomic Map