Assay Details
| Gene Symbol: KCTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198991.3 | NP_945342.1 | 2 - 3 | 120 | 128 | |
| GenBank mRNA | AF542549.1 | - | 2 - 3 | 127 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF542549.1, NM_198991.3 transcripts.
Set Membership: |
|

KCTD1
Genomic Map