Assay Details
| Gene Symbol: LOC93622 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015433.1 | 2 | 1440 | 64 | ||
| GenBank mRNA | BC006130.2 | - | 2 | 1440 | 64 | |
| BC021542.1 | - | 1 | 1716 | 64 | ||
| BC026970.1 | - | 2 | 1446 | 64 | ||
| BC040997.1 | - | 2 | 2087 | 64 | ||
| BC066911.1 | - | 1 | 1824 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23059131 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909490 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909501 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15540328 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23059141 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909513 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909533 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23059143 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909534 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909517 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909525 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23059139 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909491 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909516 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23059150 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23059138 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909518 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23059125 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909512 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23059157 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909535 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909497 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909531 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909523 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23059135 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909509 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909527 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909524 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909532 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23059134 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23059129 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909530 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909507 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909488 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909502 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909484 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P02909519 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P15540327 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23059128 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P428992 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 16460 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 24586 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 15886 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P00913839 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P428992 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_23_P428992 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909484 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909480 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059112 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059123 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059113 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909469 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059105 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059106 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059114 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909465 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909470 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909471 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909477 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059135 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059125 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059109 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909491 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059128 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059109 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059125 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909491 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909477 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909467 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909484 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909470 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059115 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909471 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059135 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059112 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909480 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909469 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909465 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059131 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059105 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059134 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059114 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P02909486 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059123 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059128 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059106 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23059113 | - | - |
| Agilent Human Genome CGH Microarray 4x44K | A_14_P133625 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P89038 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 170897 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 83835 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P16579043 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8093945 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8093944 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8093946 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8093947 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8093948 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8093949 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8093943 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 225391_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000444232.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000444232.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000444232.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000444232.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | ENST00000444232.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | NR_015433.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | NR_015433.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | NR_015433.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | NR_015433.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006695.hg.1 | NR_015433.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400146411.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400146411.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400146411.hg.1 | ENST00000444232.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400146411.hg.1 | NR_015433.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400146411.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400146411.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400146411.hg.1 | ENST00000444232.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400146411.hg.1 | NR_015433.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400146411.hg.1 | TC0400006695.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000307533.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000444232.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000444232.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000444232.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000444232.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | ENST00000444232.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | NR_015433.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | NR_015433.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | NR_015433.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | NR_015433.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400006695.hg.1 | NR_015433.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 225391_at | - | - |
| Human Genome U133 Plus 2.0 Array | 225391_PM_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225391_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225391_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225391_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225391_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225391_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225391_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225391_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225391_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225391_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225391_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 225391_at6 | - | - |
| Human Genome U95 Set | 47822_at | - | - |
| Human Genome U95 Set | 44076_PM_at | - | - |
| Human Genome U95 Set | 47822_PM_at | - | - |
| Human Genome U95 Set | 44076_at | - | - |
| Human Genome U133 Set | 225391_PM_at | - | - |
| Human Genome U133 Set | 225391_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025232 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025232 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1797181 | CR606044.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1797181 | CR616401.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1797181 | BC040997.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1797181 | BC066911.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1797181 | BC073972.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1797181 | CR606044.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1797181 | CR616401.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1797181 | BC040997.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1797181 | BC066911.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1797181 | BC073972.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1797181 | CR606044.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1797181 | CR616401.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1797181 | BC040997.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1797181 | BC066911.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1797181 | BC073972.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1797181 | CR606044.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1797181 | CR616401.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1797181 | BC040997.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1797181 | BC066911.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1797181 | BC073972.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1797181 | CR606044.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1797181 | CR616401.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1797181 | BC040997.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1797181 | BC066911.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1797181 | BC073972.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1797181 | CR606044.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1797181 | CR616401.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1797181 | BC040997.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1797181 | BC066911.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1797181 | BC073972.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1797181 | CR606044.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1797181 | CR616401.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1797181 | BC040997.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1797181 | BC066911.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1797181 | BC073972.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1797181 | CR606044.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1797181 | CR616401.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1797181 | BC040997.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1797181 | BC066911.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1797181 | BC073972.1 | human_transcriptome |

LOC93622
Genomic Map