Assay Details
| Gene Symbol: AGTPBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005251849.3 | XP_005251906.1 | 4 - 5 | 444 | 134 | |
| XM_017014546.1 | XP_016870035.1 | 4 - 5 | 407 | 134 | ||
| XM_017014547.1 | XP_016870036.1 | 4 - 5 | 444 | 134 | ||
| XM_017014549.1 | XP_016870038.1 | 4 - 5 | 444 | 134 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005251849.3, XM_017014546.1, XM_017014547.1, XM_017014549.1 transcripts.
Set Membership: |
|

AGTPBP1
Genomic Map