Assay Details
| Gene Symbol: FGFR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163213.1 | NP_001156685.1 | 8 - 9 | 1339 | 128 | |
| XM_006713868.1 | XP_006713931.1 | 8 - 9 | 1352 | 128 | ||
| XM_006713869.1 | XP_006713932.1 | 8 - 9 | 1352 | 128 | ||
| XM_006713870.1 | XP_006713933.1 | 8 - 9 | 1352 | 128 | ||
| XM_006713871.1 | XP_006713934.1 | 8 - 9 | 1352 | 128 | ||
More Information
Set Membership: |
|

FGFR3
Genomic Map