Assay Details
| Gene Symbol: IGF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | X06160.1 | - | 2 - 3 | 250 | 110 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381389.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381392.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000418738.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000418738.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000418738.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000418738.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000418738.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000430034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000430034.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381389.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381392.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381395.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000381406.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000416167.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000418738.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000418738.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000418738.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000418738.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000418738.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000430034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000430034.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | ENST00000434045.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_000612.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001127598.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291861.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291862.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013131.hg.1 | NM_001291862.2 | Refseq |

IGF2
Genomic Map