| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16014714.hg.1 |
ENST00000305596.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16014714.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16014714.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR16014714.hg.1 |
TC16001027.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000564525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000564525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000564525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000564525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000564525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000564525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000564525.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600176959.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600176959.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600176959.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600176959.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600176959.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600176959.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600176959.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600176959.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600176959.hg.1 |
TC1600009961.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000305596.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000564525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000564525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000564525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000564525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000564525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000564525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000564525.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
ENST00000569466.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_001243646.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1600009961.hg.1 |
NM_006110.2 |
Refseq |