Assay Details
| Gene Symbol: ZNF445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181489.5 | NP_852466.1 | 4 - 5 | 940 | 87 | |
| XM_005265102.2 | XP_005265159.1 | 3 - 4 | 798 | 87 | ||
| XM_011533674.2 | XP_011531976.1 | 3 - 4 | 763 | 87 | ||
| XM_017006345.1 | XP_016861834.1 | 3 - 4 | 855 | 87 | ||
| XM_017006346.1 | XP_016861835.1 | 3 - 4 | 798 | 87 | ||
| GenBank mRNA | AY262260.1 | - | 2 - 3 | 743 | 87 | |
| AY279350.1 | - | 3 - 4 | 756 | 87 | ||
| AY295873.2 | - | 4 - 5 | 940 | 87 | ||
| BC101486.1 | - | 2 - 3 | 654 | 8068992 | 87 | |
| BC143439.1 | - | 2 - 3 | 655 | 9051947 | 87 | |
| L32162.1 | - | 3 - 4 | 756 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001337.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03022879.hg.1 | TC03001337.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03022879.hg.1 | ENST00000340674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03022879.hg.1 | ENST00000396077.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03022879.hg.1 | ENST00000425708.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03022879.hg.1 | ENST00000430301.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03022879.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300179413.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300179413.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300179413.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300179413.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300179413.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300179413.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300179413.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300179413.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300179413.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300179413.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300179413.hg.1 | TC0300010889.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000396077.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000425708.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000460529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000474600.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | ENST00000617032.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010889.hg.1 | NM_181489.5 | Refseq |

ZNF445
Genomic Map