Assay Details
| Gene Symbol: UBL4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014235.4 | NP_055050.1 | 1 - 2 | 136 | 95 | |
| GenBank mRNA | AK000405.1 | - | 1 - 2 | 71 | 95 | |
| BC043346.2 | - | 1 - 2 | 63 | 6058941 | 95 | |
| BC053589.1 | - | 1 - 2 | 97 | 5246329 | 95 | |
| DA012854.1 | - | 1 - 2 | 136 | 95 | ||
| DR003955.1 | - | 1 - 2 | 76 | 95 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001535.hg
Additional Information:
This assay is in the 5'UTR region of the BC043346.2,BC053589.1,NM_014235.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020505.hg.1 | TC0X001535.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020505.hg.1 | ENST00000369653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020505.hg.1 | ENST00000369660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020505.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020506.hg.1 | TC0X001535.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020506.hg.1 | ENST00000369653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020506.hg.1 | ENST00000369660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020506.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020500.hg.1 | TC0X001535.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020500.hg.1 | ENST00000369653.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020500.hg.1 | ENST00000369660.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020500.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X020500.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000481237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000481237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000481237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000481237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000630530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | ENST00000630530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176521.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176521.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176521.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176521.hg.1 | ENST00000481237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176521.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176521.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176521.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176521.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176521.hg.1 | ENST00000481237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176521.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176521.hg.1 | TC0X00011205.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00176528.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176528.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176528.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176528.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176528.hg.1 | TC0X00011205.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000477777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000481237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000481237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000481237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000481237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000630530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | ENST00000630530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011205.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176522.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176522.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176522.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176522.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176522.hg.1 | ENST00000481237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176522.hg.1 | ENST00000630530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176522.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176522.hg.1 | ENST00000369653.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176522.hg.1 | ENST00000369660.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176522.hg.1 | ENST00000417913.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176522.hg.1 | ENST00000421431.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176522.hg.1 | ENST00000481237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176522.hg.1 | ENST00000630530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00176522.hg.1 | NM_014235.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00176522.hg.1 | TC0X00011205.hg.1 | Clariom D TC |

UBL4A
Genomic Map