Assay Details
| Gene Symbol: TNFSF14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172014.3 | NP_742011.2 | 1 - 2 | 198 | 86 | |
| GenBank mRNA | AY028261.1 | - | 1 - 2 | 157 | 86 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001102.hg.1
Set Membership: |
|

TNFSF14
Genomic Map