Assay Details
| Gene Symbol: XPNPEP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022098.3 | NP_071381.1 | 2 - 3 | 281 | 68 | |
| GenBank mRNA | AK301635.1 | - | 2 - 3 | 280 | 68 | |
| AK310502.1 | - | 2 - 3 | 539 | 68 | ||
| AK313770.1 | - | 2 - 3 | 242 | 68 | ||
| AL365514.1 | - | 2 - 3 | 264 | 68 | ||
| BC001208.1 | - | 2 - 3 | 216 | 3502251 | 68 | |
| BC001681.2 | - | 2 - 3 | 248 | 2823656 | 68 | |
| BC004989.1 | - | 2 - 3 | 248 | 2823656 | 68 | |
| BX648018.1 | - | 2 - 3 | 276 | 68 | ||
| CR456442.1 | - | 2 - 3 | 217 | 68 | ||
| CR457396.1 | - | 2 - 3 | 180 | 68 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000327.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000465258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000465258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000465258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000614001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000614001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | ENST00000614001.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006314.hg.1 | ENST00000357137.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006314.hg.1 | ENST00000414396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006314.hg.1 | ENST00000541156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006314.hg.1 | ENST00000544094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006314.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006314.hg.1 | TC22000327.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006313.hg.1 | ENST00000357137.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006313.hg.1 | ENST00000414396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006313.hg.1 | ENST00000541156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006313.hg.1 | ENST00000544094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006313.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006313.hg.1 | TC22000327.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200169083.hg.1 | TC2200009277.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169083.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169083.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169083.hg.1 | ENST00000465258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169083.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169083.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169083.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169083.hg.1 | ENST00000465258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169083.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169084.hg.1 | TC2200009277.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200169084.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169084.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169084.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200169084.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169084.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200169084.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000357137.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000417688.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000428799.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000465258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000465258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000465258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000482652.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000614001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000614001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | ENST00000614001.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_001204827.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200009277.hg.1 | NM_022098.3 | Refseq |

XPNPEP3
Genomic Map