Assay Details
| Gene Symbol: DOCK9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005254034.3 | XP_005254091.1 | 37 - 38 | 4110 | 95 | |
| XM_006719922.1 | XP_006719985.1 | 37 - 38 | 4271 | 95 | ||
| XM_006719924.2 | XP_006719987.1 | 37 - 38 | 4110 | 95 | ||
| XM_006719925.1 | XP_006719988.1 | 37 - 38 | 4238 | 95 | ||
| XM_017020503.1 | XP_016875992.1 | 37 - 38 | 4111 | 95 | ||
| XM_017020504.1 | XP_016875993.1 | 37 - 38 | 4111 | 95 | ||
| XM_017020505.1 | XP_016875994.1 | 37 - 38 | 4111 | 95 | ||
| XM_017020507.1 | XP_016875996.1 | 37 - 38 | 4111 | 95 | ||
| XM_017020509.1 | XP_016875998.1 | 37 - 38 | 4238 | 95 | ||
| XM_017020512.1 | XP_016876001.1 | 37 - 38 | 4238 | 95 | ||
| XM_017020514.1 | XP_016876003.1 | 37 - 38 | 4238 | 95 | ||
| XM_017020516.1 | XP_016876005.1 | 37 - 38 | 4238 | 95 | ||
| GenBank mRNA | AK300298.1 | - | 2 - 3 | 123 | 95 | |
| AK300331.1 | - | 3 - 4 | 209 | 95 | ||
More Information
Set Membership: |
|

DOCK9
Genomic Map