Assay Details
| Gene Symbol: NRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005267708.4 | XP_005267765.1 | 1 - 2 | 199 | 121 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005267708.4 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
regulation of rhodopsin gene expression
visual perception
positive regulation of rhodopsin gene expression
positive regulation of transcription from RNA polymerase II promoter
retinal rod cell development
response to stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
protein binding
leucine zipper domain binding
transcription from RNA polymerase II promoter
regulation of rhodopsin gene expression
visual perception
positive regulation of rhodopsin gene expression
positive regulation of transcription from RNA polymerase II promoter
retinal rod cell development
response to stimulus
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
protein binding
leucine zipper domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000396997.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000397002.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000558280.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000560550.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | ENST00000561028.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400008739.hg.1 | NM_006177.4 | Refseq |

NRL
Genomic Map