Assay Details
| Gene Symbol: ALDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000690.3 | NP_000681.2 | 6 - 7 | 776 | 72 | |
| NM_001204889.1 | NP_001191818.1 | 5 - 6 | 635 | 72 | ||
| GenBank mRNA | AK223373.1 | - | 6 - 7 | 718 | 72 | |
| AK301375.1 | - | 5 - 6 | 573 | 72 | ||
| AK314856.1 | - | 6 - 7 | 698 | 72 | ||
| AY621070.1 | - | 6 - 7 | 741 | 72 | ||
| BC002967.1 | - | 6 - 7 | 726 | 3543343 | 72 | |
| BC071839.1 | - | 6 - 7 | 685 | 5477768 | 72 | |
| CR456991.1 | - | 6 - 7 | 680 | 72 | ||
| K03001.1 | - | 3 - 4 | 326 | 72 | ||
| M26760.1 | - | 3 - 4 | 326 | 72 | ||
| M54931.1 | - | 1 - 2 | 41 | 72 | ||
| X05409.1 | - | 6 - 7 | 713 | 72 | ||
| Y00109.1 | - | 6 - 7 | 704 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12003212.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000551450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | ENST00000551450.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011479.hg.1 | TC12003212.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011479.hg.1 | ENST00000261733.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011479.hg.1 | ENST00000416293.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011479.hg.1 | ENST00000551906.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011479.hg.1 | ENST00000552234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011479.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12011479.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000549106.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000551450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | ENST00000551450.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012707.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201647.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201647.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201647.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201647.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201647.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201647.hg.1 | ENST00000261733.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201647.hg.1 | ENST00000416293.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201647.hg.1 | ENST00000548536.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201647.hg.1 | NM_000690.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201647.hg.1 | NM_001204889.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201647.hg.1 | TC1200012707.hg.1 | Clariom D TC |

ALDH2
Genomic Map