Assay Details
| Gene Symbol: TAF6L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006473.3 | NP_006464.1 | 8 - 9 | 1030 | 121 | |
| XM_005273714.1 | XP_005273771.1 | 8 - 9 | 895 | 121 | ||
| GenBank mRNA | AF069735.1 | - | 7 - 8 | 829 | 121 | |
| AK289499.1 | - | 8 - 9 | 940 | 121 | ||
| AK314330.1 | - | 8 - 9 | 916 | 121 | ||
| BC008785.2 | - | 8 - 9 | 930 | 121 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000564.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin remodeling
DNA-templated transcription, initiation
regulation of transcription from RNA polymerase II promoter
histone H3 acetylation
regulation of sequence-specific DNA binding transcription factor activity
DNA binding
transcription coactivator activity
histone acetyltransferase activity
protein binding
protein heterodimerization activity
chromatin remodeling
DNA-templated transcription, initiation
regulation of transcription from RNA polymerase II promoter
histone H3 acetylation
regulation of sequence-specific DNA binding transcription factor activity
DNA binding
transcription coactivator activity
histone acetyltransferase activity
protein binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11006810.hg.1 | ENST00000294168.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006810.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11006810.hg.1 | TC11000564.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000525405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000525405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000525405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000525405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000525405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000525405.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000526261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000526261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000526261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000529509.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000529509.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000529509.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000529509.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000529509.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000531265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000531265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000531265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000531265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000531265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000532915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000532915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000532915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000532915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000532915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000532915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000532915.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000533823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000533823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000533823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | ENST00000533823.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100156109.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156109.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100156109.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100156109.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100156109.hg.1 | TC1100007875.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000294168.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000524976.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000525405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000525405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000525405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000525405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000525405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000525405.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000526261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000526261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000526261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000529509.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000529509.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000529509.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000529509.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000529509.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000531265.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000531265.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000531265.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000531265.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000531265.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000532915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000532915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000532915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000532915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000532915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000532915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000532915.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000533823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000533823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000533823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | ENST00000533823.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | NM_006473.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100007875.hg.1 | NM_006473.3 | Refseq |

TAF6L
Genomic Map