Assay Details
| Gene Symbol: TUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022830.2 | NP_073741.2 | 3 - 4 | 739 | 87 | |
| GenBank mRNA | AB101208.1 | - | 3 - 4 | 653 | 87 | |
| AK026000.1 | - | 3 - 4 | 646 | 87 | ||
| AK292610.1 | - | 3 - 4 | 738 | 87 | ||
| BC110910.1 | - | 3 - 4 | 628 | 6141937 | 87 | |
| BC128263.1 | - | 3 - 4 | 632 | 40118672 | 87 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11003484.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mRNA polyadenylation
snRNA processing
histone mRNA catabolic process
pre-mRNA cleavage required for polyadenylation
RNA binding
mRNA 3'-UTR binding
polynucleotide adenylyltransferase activity
protein binding
ATP binding
enzyme binding
metal ion binding
RNA uridylyltransferase activity
mRNA polyadenylation
snRNA processing
histone mRNA catabolic process
pre-mRNA cleavage required for polyadenylation
RNA binding
mRNA 3'-UTR binding
polynucleotide adenylyltransferase activity
protein binding
ATP binding
enzyme binding
metal ion binding
RNA uridylyltransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11021686.hg.1 | TC11003484.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021686.hg.1 | ENST00000308436.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021686.hg.1 | ENST00000476907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021686.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000278279.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000278279.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000278279.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000463241.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000463241.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000463241.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000463241.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000463241.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000463241.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000469480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000469480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000469480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000469480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000469480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000469480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000469480.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000478537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000478537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000478537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000478537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000494385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000494385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000494385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000494385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | ENST00000494385.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021687.hg.1 | TC11003484.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021687.hg.1 | ENST00000308436.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021687.hg.1 | ENST00000476907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11021687.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000278279.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000278279.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000278279.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000463241.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000463241.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000463241.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000463241.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000463241.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000463241.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000469480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000469480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000469480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000469480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000469480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000469480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000469480.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000478537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000478537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000478537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000478537.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000494385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000494385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000494385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000494385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | ENST00000494385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013171.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204256.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204256.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204256.hg.1 | ENST00000494385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204256.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204256.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204256.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204256.hg.1 | ENST00000494385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204256.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204256.hg.1 | TC1100013171.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100204255.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204255.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204255.hg.1 | ENST00000494385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204255.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204255.hg.1 | ENST00000308436.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204255.hg.1 | ENST00000476907.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204255.hg.1 | ENST00000494385.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100204255.hg.1 | NM_022830.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100204255.hg.1 | TC1100013171.hg.1 | Clariom D TC |

TUT1
Genomic Map