Assay Details
Gene Symbol: UBOX5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038395.1 | 1 | 92 | 49 | ||
GenBank mRNA | AK097516.1 | - | 1 | 92 | 49 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000034.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ArrayStar
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR20000569.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000569.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20000569.hg.1 | TC20000034.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20000572.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000572.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20000572.hg.1 | TC20000034.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20000565.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000565.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20000565.hg.1 | TC20000034.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000454019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000454019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | ENST00000454019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20000566.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000566.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20000566.hg.1 | TC20000034.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20000568.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000568.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20000568.hg.1 | TC20000034.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20000571.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000571.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20000571.hg.1 | TC20000034.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR20000564.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20000564.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20000564.hg.1 | TC20000034.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2000144948.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144948.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144948.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144948.hg.1 | TC2000006538.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000144948.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144948.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144948.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144942.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144942.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144942.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144942.hg.1 | TC2000006538.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000144942.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144942.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144942.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144945.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144945.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144945.hg.1 | ENST00000454019.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144945.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144945.hg.1 | TC2000006538.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000144945.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144945.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144945.hg.1 | ENST00000454019.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144945.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144940.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144940.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144940.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144940.hg.1 | TC2000006538.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000144940.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144940.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144940.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144941.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144941.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144941.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144941.hg.1 | TC2000006538.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000144941.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144941.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144941.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144943.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144943.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144943.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144943.hg.1 | TC2000006538.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000144943.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144943.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144943.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144946.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144946.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144946.hg.1 | ENST00000454019.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144946.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144946.hg.1 | TC2000006538.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000144946.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144946.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144946.hg.1 | ENST00000454019.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144946.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144949.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144949.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144949.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000144949.hg.1 | TC2000006538.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000144949.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144949.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000144949.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000446537.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000454019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000454019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | ENST00000454019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000006538.hg.1 | NR_038395.1 | Refseq |
Human Genome U95 Set | 77585_PM_at | - | - |
Human Genome U95 Set | 77589_at | - | - |
Human Genome U95 Set | 77587_PM_g_at | - | - |
Human Genome U95 Set | 77589_PM_at | - | - |
Human Genome U95 Set | 77587_g_at | - | - |
Human Genome U95 Set | 77585_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017990 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A017990 | - | - |
