Assay Details
| Gene Symbol: DNLZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073565.1 | 1 - 2 | 328 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186821.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186821.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186821.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186821.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186821.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186821.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186821.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186821.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186821.hg.1 | TC0900012284.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | ENST00000371739.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900012284.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186823.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186823.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186823.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186823.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186823.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186823.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186823.hg.1 | TC0900012284.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900186819.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186819.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186819.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186819.hg.1 | ENST00000371738.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900186819.hg.1 | NM_001080849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186819.hg.1 | NR_073565.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900186819.hg.1 | TC0900012284.hg.1 | Clariom D TC |

DNLZ
Genomic Map