Assay Details
| Gene Symbol: DNLZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080849.2 | NP_001074318.1 | 2 - 3 | 466 | 81 | |
| NR_073565.1 | 2 - 3 | 470 | 81 | |||
| GenBank mRNA | AI435052.1 | - | 1 - 2 | 279 | 81 | |
| BC146915.1 | - | 2 - 3 | 466 | 9056843 | 81 | |
| BC146924.1 | - | 2 - 3 | 466 | 9056852 | 81 | |
More Information
Set Membership: |
|

DNLZ
Genomic Map