Assay Details
| Gene Symbol: SEMA3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152754.2 | NP_689967.2 | 11 - 12 | 1452 | 70 | |
| XM_011515960.1 | XP_011514262.1 | 13 - 14 | 1762 | 70 | ||
| XM_011515961.2 | XP_011514263.1 | 11 - 12 | 1287 | 70 | ||
| XM_017011873.1 | XP_016867362.1 | 12 - 13 | 1951 | 70 | ||
| GenBank mRNA | AY358937.1 | - | 12 - 13 | 1518 | 70 | |
| BC029590.1 | - | 4 - 5 | 538 | 5272175 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001570.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07023717.hg.1 | TC07001570.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07023717.hg.1 | ENST00000284136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07023717.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07023717.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700183336.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183336.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183336.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700183336.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183336.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183336.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700183336.hg.1 | TC0700011675.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |

SEMA3D
Genomic Map