Assay Details
| Gene Symbol: SEMA3D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152754.2 | NP_689967.2 | 4 - 5 | 528 | 105 | |
| XM_011515960.1 | XP_011514262.1 | 6 - 7 | 838 | 105 | ||
| XM_011515961.2 | XP_011514263.1 | 4 - 5 | 363 | 105 | ||
| XM_017011873.1 | XP_016867362.1 | 5 - 6 | 1027 | 105 | ||
| GenBank mRNA | AK309116.1 | - | 6 - 7 | 838 | 105 | |
| AY358937.1 | - | 5 - 6 | 594 | 105 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001570.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011515961.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07023729.hg.1 | TC07001570.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07023729.hg.1 | ENST00000284136.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07023729.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07023729.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000463315.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | ENST00000484038.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011675.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700183348.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183348.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183348.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700183348.hg.1 | ENST00000284136.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183348.hg.1 | ENST00000444867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700183348.hg.1 | NM_152754.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700183348.hg.1 | TC0700011675.hg.1 | Clariom D TC |

SEMA3D
Genomic Map