Assay Details
Gene Symbol: SHTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127211.2 | NP_001120683.1 | 16 - 17 | 2170 | 151 | |
NM_001258298.1 | NP_001245227.1 | 15 - 16 | 2117 | 151 | ||
XM_017016462.1 | XP_016871951.1 | 16 - 17 | 1680 | 151 | ||
GenBank mRNA | AK001984.1 | - | 2 - 3 | 264 | 151 | |
AK091381.1 | - | 6 - 7 | 803 | 151 | ||
AK091578.1 | - | 15 - 16 | 1876 | 151 | ||
AK095419.1 | - | 6 - 7 | 629 | 151 | ||
AK304865.1 | - | 15 - 16 | 1764 | 151 | ||
BC022348.1 | - | 5 - 6 | 512 | 151 | ||
CR749417.1 | - | 16 - 17 | 1778 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
substrate-dependent cell migration, cell extension
Ras protein signal transduction
axonogenesis
Cdc42 protein signal transduction
netrin-activated signaling pathway
positive regulation of axon extension
neuron projection morphogenesis
cytoplasmic actin-based contraction involved in cell motility
endoplasmic reticulum polarization
actin filament bundle retrograde transport
cell-cell adhesion
regulation of establishment of cell polarity
positive regulation of neuron migration
protein binding
kinesin binding
actin filament binding
cadherin binding involved in cell-cell adhesion
substrate-dependent cell migration, cell extension
Ras protein signal transduction
axonogenesis
Cdc42 protein signal transduction
netrin-activated signaling pathway
positive regulation of axon extension
neuron projection morphogenesis
cytoplasmic actin-based contraction involved in cell motility
endoplasmic reticulum polarization
actin filament bundle retrograde transport
cell-cell adhesion
regulation of establishment of cell polarity
positive regulation of neuron migration
protein binding
kinesin binding
actin filament binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000401506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000401506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000401506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000401506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000401506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000401506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000401506.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000406087.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000406087.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000406087.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000406087.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000406087.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000406087.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000407930.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000407930.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000407930.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000407930.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000407930.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000407930.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000412933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000412933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000412933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000412933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000412933.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000464412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000464412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000464412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000464412.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000484150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000484150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000484150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000484150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000484150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | ENST00000484150.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000395323.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000401506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000401506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000401506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000401506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000401506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000401506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000401506.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000406087.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000406087.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000406087.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000406087.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000406087.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000406087.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000407930.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000407930.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000407930.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000407930.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000407930.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000407930.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000412933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000412933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000412933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000412933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000412933.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000464412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000464412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000464412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000464412.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000467242.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000484150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000484150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000484150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000484150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000484150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | ENST00000484150.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | NM_030915.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200016424.hg.1 | NM_030915.3 | Refseq |
