Assay Details
| Gene Symbol: COL24A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152890.5 | NP_690850.2 | 53 - 54 | 4419 | 62 | |
| XM_017000924.1 | XP_016856413.1 | 55 - 56 | 4592 | 62 | ||
| XM_017000925.1 | XP_016856414.1 | 55 - 56 | 4547 | 62 | ||
| XM_017000926.1 | XP_016856415.1 | 54 - 55 | 4789 | 62 | ||
| XM_017000927.1 | XP_016856416.1 | 53 - 54 | 4735 | 62 | ||
| XM_017000928.1 | XP_016856417.1 | 36 - 37 | 3501 | 62 | ||
| XM_017000929.1 | XP_016856418.1 | 35 - 36 | 3455 | 62 | ||
| GenBank mRNA | AY244357.1 | - | 53 - 54 | 4409 | 62 | |
| BC113654.1 | - | 53 - 54 | 4416 | 8322706 | 62 | |
| BC117170.1 | - | 53 - 54 | 4416 | 40125721 | 62 | |
| BC143715.1 | - | 53 - 54 | 4388 | 62 | ||
| BC143717.1 | - | 53 - 54 | 4412 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002832.hg.1
Set Membership: |
|

COL24A1
Genomic Map