Assay Details
| Gene Symbol: COL24A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152890.5 | NP_690850.2 | 54 - 55 | 4453 | 139 | |
| XM_017000924.1 | XP_016856413.1 | 56 - 57 | 4626 | 139 | ||
| XM_017000925.1 | XP_016856414.1 | 56 - 57 | 4581 | 139 | ||
| XM_017000926.1 | XP_016856415.1 | 55 - 56 | 4823 | 139 | ||
| XM_017000927.1 | XP_016856416.1 | 54 - 55 | 4769 | 139 | ||
| XM_017000928.1 | XP_016856417.1 | 37 - 38 | 3535 | 139 | ||
| XM_017000929.1 | XP_016856418.1 | 36 - 37 | 3489 | 139 | ||
| GenBank mRNA | AY244357.1 | - | 54 - 55 | 4443 | 139 | |
| BC113654.1 | - | 54 - 55 | 4450 | 8322706 | 139 | |
| BC117170.1 | - | 54 - 55 | 4450 | 40125721 | 139 | |
| BC143715.1 | - | 54 - 55 | 4422 | 139 | ||
| BC143717.1 | - | 54 - 55 | 4446 | 139 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002832.hg.1
Set Membership: |
|

COL24A1
Genomic Map