Assay Details
| Gene Symbol: MAP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198611.1 | NP_001185540.1 | 5 - 6 | 843 | 87 | |
| NM_001198616.1 | NP_001185545.1 | 5 - 6 | 939 | 87 | ||
| XM_011536244.2 | XP_011534546.1 | 5 - 6 | 1754 | 87 | ||
| XM_011536245.1 | XP_011534547.1 | 5 - 6 | 633 | 87 | ||
| XM_011536246.2 | XP_011534548.1 | 5 - 6 | 1735 | 87 | ||
| XM_017011471.1 | XP_016866960.1 | 6 - 7 | 1364 | 87 | ||
| GenBank mRNA | AJ242502.1 | - | 5 - 6 | 528 | 87 | |
| AK294461.1 | - | 5 - 6 | 613 | 87 | ||
More Information
Set Membership: |
|

MAP7
Genomic Map