Assay Details
| Gene Symbol: TMEM57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282564.1 | NP_001269493.1 | 5 - 6 | 866 | 91 | |
| NM_018202.5 | NP_060672.2 | 7 - 8 | 1547 | 91 | ||
| XM_005245931.2 | XP_005245988.1 | 7 - 8 | 1508 | 91 | ||
| XM_011541704.2 | XP_011540006.1 | 7 - 8 | 1581 | 91 | ||
| GenBank mRNA | AK001609.1 | - | 7 - 8 | 1458 | 91 | |
| AY845015.1 | - | 7 - 8 | 1477 | 91 | ||
| AY845035.1 | - | 5 - 6 | 734 | 91 | ||
| AY845036.1 | - | 2 - 3 | 341 | 91 | ||
| BC032427.1 | - | 7 - 8 | 1415 | 5246967 | 91 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000322.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000470035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000470035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000470035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | ENST00000470035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004917.hg.1 | TC01000322.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004917.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004917.hg.1 | ENST00000399763.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004917.hg.1 | ENST00000399766.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01004917.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151584.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151584.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151584.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151584.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151584.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151584.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100151584.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151584.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100151584.hg.1 | TC0100007403.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000374343.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000399766.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000470035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000470035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000470035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | ENST00000470035.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_001282564.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007403.hg.1 | NM_018202.5 | Refseq |

TMEM57
Genomic Map