Assay Details
| Gene Symbol: ADAMTS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139025.4 | NP_620594.1 | 8 - 9 | 1426 | 123 | |
| NM_139027.4 | NP_620596.2 | 8 - 9 | 1426 | 123 | ||
| XM_017014232.1 | XP_016869721.1 | 8 - 9 | 1637 | 123 | ||
| XM_017014233.1 | XP_016869722.1 | 8 - 9 | 1038 | 123 | ||
| XM_017014235.1 | XP_016869724.1 | 8 - 9 | 2207 | 123 | ||
| GenBank mRNA | AB069698.2 | - | 8 - 9 | 1426 | 123 | |
| AF414401.1 | - | 8 - 9 | 1043 | 123 | ||
| AJ420810.1 | - | 8 - 9 | 1102 | 123 | ||
| AY055376.1 | - | 8 - 9 | 1090 | 123 | ||
| AY358118.1 | - | 1 - 2 | 194 | 123 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AY358118.1 transcripts.
Set Membership: |
|

ADAMTS13
Genomic Map