Assay Details
| Gene Symbol: TUSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007275.2 | NP_009206.1 | 3 | 468 | 255 | |
| GenBank mRNA | AF055479.1 | - | 3 | 476 | 255 | |
| AK124058.1 | - | 4 | 374 | 227 | ||
| BC023976.2 | - | 3 | 420 | 3504783 | 255 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03003330.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
natural killer cell differentiation
phagocytosis
inflammatory response
cell cycle
cell-cell signaling
cell proliferation
interleukin-15 production
negative regulation of interleukin-17 production
positive regulation of interleukin-10 production
cell maturation
regulation of mitochondrial membrane potential
response to defense-related host reactive oxygen species production
neutrophil mediated killing of gram-negative bacterium
chemokine (C-C motif) ligand 5 production
regulation of reactive oxygen species metabolic process
protein binding
natural killer cell differentiation
phagocytosis
inflammatory response
cell cycle
cell-cell signaling
cell proliferation
interleukin-15 production
negative regulation of interleukin-17 production
positive regulation of interleukin-10 production
cell maturation
regulation of mitochondrial membrane potential
response to defense-related host reactive oxygen species production
neutrophil mediated killing of gram-negative bacterium
chemokine (C-C motif) ligand 5 production
regulation of reactive oxygen species metabolic process
protein binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000417867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000417867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000417867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000417867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000417867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000463304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000463304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000463304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000463304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000463304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | ENST00000463304.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025702.hg.1 | TC03003330.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025702.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025702.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025703.hg.1 | TC03003330.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025703.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03025703.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207801.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207801.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207801.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207801.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207801.hg.1 | TC0300013985.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300207803.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207803.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207803.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207803.hg.1 | ENST00000463304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207803.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207803.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207803.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207803.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207803.hg.1 | ENST00000463304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207803.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207803.hg.1 | TC0300013985.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300207802.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207802.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207802.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207802.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207802.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207802.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207802.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300207802.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300207802.hg.1 | TC0300013985.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000232496.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000417867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000417867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000417867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000417867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000417867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000421918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000454201.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000463304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000463304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000463304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000463304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000463304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | ENST00000463304.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013985.hg.1 | NM_007275.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 189081 | - | - |

TUSC2
Genomic Map