| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000371539.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000371539.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000371539.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000371539.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000371539.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000371539.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000371539.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000395997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000395997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000395997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000395997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000395997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000395997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000395997.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000481363.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000481363.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000481363.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000483130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000483130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000483130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000483130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
ENST00000483130.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000217086.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000371539.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000371539.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000371539.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000371539.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000371539.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000371539.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000371539.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000395997.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000395997.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000395997.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000395997.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000395997.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000395997.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000395997.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000481363.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000481363.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000481363.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000483130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000483130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000483130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000483130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
ENST00000483130.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009462.hg.1 |
NM_020436.4 |
Refseq |