Assay Details
| Gene Symbol: HAO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016527.3 | NP_057611.1 | 1 - 2 | 133 | 75 | |
| GenBank mRNA | AF231917.1 | - | 1 - 2 | 71 | 75 | |
| AY513277.1 | - | 1 - 2 | 38 | 75 | ||
| CB121424.1 | - | 1 - 2 | 130 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF231917.1, AY513277.1, NM_016527.3 transcripts.
Set Membership: |
|

HAO2
Genomic Map