Assay Details
| Gene Symbol: FLVCR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014053.3 | NP_054772.1 | 4 - 5 | 1295 | 91 | |
| XM_011509446.2 | XP_011507748.1 | 4 - 5 | 1271 | 91 | ||
| GenBank mRNA | AF118637.1 | - | 4 - 5 | 1295 | 91 | |
| AK001419.1 | - | 4 - 5 | 472 | 91 | ||
| AK314481.1 | - | 4 - 5 | 1271 | 91 | ||
| BC048312.1 | - | 4 - 5 | 1233 | 4417876 | 91 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001781.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel development
in utero embryonic development
transport
mitochondrial transport
cellular iron ion homeostasis
multicellular organism development
heme transport
erythrocyte differentiation
multicellular organism growth
embryonic digit morphogenesis
erythrocyte maturation
regulation of organ growth
spleen development
embryonic skeletal system morphogenesis
transmembrane transport
head morphogenesis
heme export
transporter activity
protein binding
heme transporter activity
blood vessel development
in utero embryonic development
transport
mitochondrial transport
cellular iron ion homeostasis
multicellular organism development
heme transport
erythrocyte differentiation
multicellular organism growth
embryonic digit morphogenesis
erythrocyte maturation
regulation of organ growth
spleen development
embryonic skeletal system morphogenesis
transmembrane transport
head morphogenesis
heme export
transporter activity
protein binding
heme transporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01027847.hg.1 | TC01001781.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027847.hg.1 | ENST00000366971.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027847.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027847.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027848.hg.1 | TC01001781.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027848.hg.1 | ENST00000366971.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027848.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01027848.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000579295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000579295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000579295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | ENST00000579295.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185055.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185055.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185055.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185055.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185055.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185055.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185055.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185055.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185055.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185055.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185055.hg.1 | TC0100011549.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100185057.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185057.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185057.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185057.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185057.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185057.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185057.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185057.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185057.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185057.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185057.hg.1 | TC0100011549.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000366971.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000419102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000474693.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000483790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000579295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000579295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000579295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | ENST00000579295.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011549.hg.1 | NM_014053.3 | Refseq |

FLVCR1
Genomic Map