Assay Details
| Gene Symbol: TBC1D20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144628.3 | NP_653229.1 | 2 - 3 | 403 | 83 | |
| NR_111901.1 | 2 - 3 | 403 | 83 | |||
| XM_005260661.1 | XP_005260718.1 | 2 - 3 | 393 | 83 | ||
| GenBank mRNA | AB449906.1 | - | 2 - 3 | 255 | 83 | |
| AK095110.1 | - | 2 - 3 | 395 | 83 | ||
| AK127062.1 | - | 2 - 3 | 383 | 83 | ||
| AK291648.1 | - | 2 - 3 | 366 | 83 | ||
| BC014983.1 | - | 2 - 3 | 353 | 4861869 | 83 | |
| DA238706.1 | - | 2 - 3 | 383 | 83 | ||
| DA311494.1 | - | 2 - 3 | 395 | 83 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000541.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
acrosome assembly
ER to Golgi vesicle-mediated transport
Golgi organization
virion assembly
lipid particle organization
positive regulation of GTPase activity
positive regulation by host of viral genome replication
positive regulation by virus of viral protein levels in host cell
COPII vesicle coating
lens fiber cell morphogenesis
seminiferous tubule development
cargo loading into COPII-coated vesicle
regulation of cilium assembly
positive regulation of ER to Golgi vesicle-mediated transport
GTPase activator activity
protein binding
Rab GTPase binding
acrosome assembly
ER to Golgi vesicle-mediated transport
Golgi organization
virion assembly
lipid particle organization
positive regulation of GTPase activity
positive regulation by host of viral genome replication
positive regulation by virus of viral protein levels in host cell
COPII vesicle coating
lens fiber cell morphogenesis
seminiferous tubule development
cargo loading into COPII-coated vesicle
regulation of cilium assembly
positive regulation of ER to Golgi vesicle-mediated transport
GTPase activator activity
protein binding
Rab GTPase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20007789.hg.1 | ENST00000246077.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20007789.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20007789.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20007789.hg.1 | TC20000541.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461188.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000008130.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157068.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157068.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157068.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157068.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157068.hg.1 | NR_111901.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157068.hg.1 | TC2000008130.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000157068.hg.1 | ENST00000354200.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157068.hg.1 | ENST00000461304.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157068.hg.1 | ENST00000494633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000157068.hg.1 | NM_144628.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000157068.hg.1 | NR_111901.1 | Refseq |

TBC1D20
Genomic Map