| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009302.hg.1 |
TC06000777.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009302.hg.1 |
ENST00000257776.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06009302.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008678.hg.1 |
ENST00000257776.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008678.hg.1 |
ENST00000257776.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008678.hg.1 |
ENST00000257776.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008678.hg.1 |
ENST00000257776.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008678.hg.1 |
ENST00000257776.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008678.hg.1 |
ENST00000257776.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008678.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008678.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008678.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008678.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008678.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008678.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008678.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008678.hg.1 |
ENST00000257776.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008678.hg.1 |
ENST00000257776.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008678.hg.1 |
ENST00000257776.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008678.hg.1 |
ENST00000257776.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008678.hg.1 |
ENST00000257776.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008678.hg.1 |
ENST00000257776.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008678.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008678.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008678.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008678.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008678.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008678.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008678.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600160260.hg.1 |
ENST00000257776.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600160260.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600160260.hg.1 |
ENST00000257776.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600160260.hg.1 |
NM_138409.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600160260.hg.1 |
TC0600008678.hg.1 |
Clariom D TC |