Assay Details
| Gene Symbol: INTS6L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005262394.2 | XP_005262451.1 | 11 - 12 | 1761 | 128 | |
| XM_006724740.2 | XP_006724803.1 | 11 - 12 | 1761 | 128 | ||
| XM_006724741.3 | XP_006724804.1 | 11 - 12 | 1761 | 128 | ||
| XM_017029346.1 | XP_016884835.1 | 11 - 12 | 1761 | 128 | ||
More Information
Set Membership: |
|

INTS6L
Genomic Map