Assay Details
| Gene Symbol: PIK3R4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014602.2 | NP_055417.1 | 11 - 12 | 3303 | 119 | |
| GenBank mRNA | AK314219.1 | - | 11 - 12 | 3250 | 119 | |
| BC110318.1 | - | 11 - 12 | 3249 | 5492292 | 119 | |
| BC127105.1 | - | 11 - 12 | 3120 | 40124707 | 119 | |
| BC127106.1 | - | 11 - 12 | 3120 | 40124708 | 119 | |
| Y08991.1 | - | 11 - 12 | 3289 | 119 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001778.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
protein targeting to vacuole
phosphatidylinositol biosynthetic process
macroautophagy
pexophagy
regulation of cytokinesis
receptor catabolic process
toll-like receptor 9 signaling pathway
phosphatidylinositol-3-phosphate biosynthetic process
cellular response to glucose starvation
positive regulation of phosphatidylinositol 3-kinase activity
late endosome to vacuole transport
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
1-phosphatidylinositol-3-kinase activity
protein phosphorylation
protein targeting to vacuole
phosphatidylinositol biosynthetic process
macroautophagy
pexophagy
regulation of cytokinesis
receptor catabolic process
toll-like receptor 9 signaling pathway
phosphatidylinositol-3-phosphate biosynthetic process
cellular response to glucose starvation
positive regulation of phosphatidylinositol 3-kinase activity
late endosome to vacuole transport
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
1-phosphatidylinositol-3-kinase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03032241.hg.1 | TC03001778.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032241.hg.1 | ENST00000356763.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032241.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000504834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000504834.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000507330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000507330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000507330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000507330.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000508273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000508273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000508273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000508273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000508273.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000511760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000511760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000511760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000511760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000511760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512362.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | ENST00000512677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000504834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000504834.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000507330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000507330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000507330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000507330.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000508273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000508273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000508273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000508273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000508273.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000511760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000511760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000511760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000511760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000511760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512362.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | ENST00000512677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012426.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300192936.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300192936.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300192936.hg.1 | ENST00000356763.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300192936.hg.1 | NM_014602.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300192936.hg.1 | TC0300012426.hg.1 | Clariom D TC |

PIK3R4
Genomic Map