Assay Details
| Gene Symbol: RBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002899.3 | NP_002890.2 | 3 - 4 | 652 | 60 | |
| GenBank mRNA | AK290315.1 | - | 3 - 4 | 459 | 60 | |
| BC121052.2 | - | 3 - 4 | 410 | 40118103 | 60 | |
| BU734286.1 | - | 1 - 2 | 60 | |||
| CR541979.1 | - | 3 - 4 | 355 | 60 | ||
| CR542005.1 | - | 3 - 4 | 355 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001834.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000483943.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000483943.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000483943.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000483943.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000483943.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000483943.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000483943.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000487424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000487424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000487424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000487424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000487424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000487424.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000492918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000492918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000492918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000492918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000492918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000492918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000492918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000617459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000617459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000617459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000617459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000617459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000617459.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000619087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000619087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000619087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000619087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | ENST00000619087.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_001130992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_001130992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_001130992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_001130992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_001130992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_001130992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_001130992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_001130993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_001130993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_001130993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_001130993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_001130993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_001130993.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033186.hg.1 | TC03001834.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033186.hg.1 | ENST00000232219.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033186.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033187.hg.1 | TC03001834.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033187.hg.1 | ENST00000232219.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03033187.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000483943.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000483943.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000483943.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000483943.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000483943.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000483943.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000483943.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000487424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000487424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000487424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000487424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000487424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000487424.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000492918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000492918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000492918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000492918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000492918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000492918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000492918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000617459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000617459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000617459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000617459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000617459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000617459.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000619087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000619087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000619087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000619087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | ENST00000619087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_001130992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_001130992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_001130992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_001130992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_001130992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_001130992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_001130992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_001130993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_001130993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_001130993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_001130993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_001130993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_001130993.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012598.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194237.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194237.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194237.hg.1 | ENST00000619087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194237.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194237.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194237.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194237.hg.1 | ENST00000619087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194237.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194237.hg.1 | TC0300012598.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300194236.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194236.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194236.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194236.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194236.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194236.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194236.hg.1 | TC0300012598.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300194238.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194238.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194238.hg.1 | ENST00000619087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194238.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194238.hg.1 | ENST00000232219.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194238.hg.1 | ENST00000474490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194238.hg.1 | ENST00000619087.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300194238.hg.1 | NM_002899.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300194238.hg.1 | TC0300012598.hg.1 | Clariom D TC |

RBP1
Genomic Map