Assay Details
| Gene Symbol: CXCL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002993.3 | NP_002984.1 | 3 - 4 | 516 | 101 | |
| GenBank mRNA | BC013744.1 | - | 3 - 4 | 351 | 4471095 | 101 |
| U81234.1 | - | 3 - 4 | 384 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

CXCL6
Genomic Map