Assay Details
| Gene Symbol: DDX51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175066.3 | NP_778236.2 | 12 - 13 | 1817 | 129 | |
| GenBank mRNA | AB593157.1 | - | 12 - 13 | 1837 | 129 | |
| AK097078.1 | - | 12 - 13 | 1817 | 129 | ||
| BC012461.2 | - | 3 - 4 | 340 | 129 | ||
| BC040185.2 | - | 12 - 13 | 1790 | 4801253 | 129 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12002148.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12028121.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028121.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12028121.hg.1 | TC12002148.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000545991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | ENST00000546058.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012512.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197632.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197632.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197632.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197632.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197632.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197632.hg.1 | ENST00000329073.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197632.hg.1 | ENST00000397333.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197632.hg.1 | ENST00000462829.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197632.hg.1 | ENST00000541489.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200197632.hg.1 | NM_175066.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200197632.hg.1 | TC1200012512.hg.1 | Clariom D TC |

DDX51
Genomic Map