Assay Details
| Gene Symbol: FILIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300866.1 | NP_001287795.1 | 1 - 2 | 417 | 99 | |
| NM_015687.3 | NP_056502.1 | 1 - 2 | 417 | 99 | ||
| XM_005248713.3 | XP_005248770.1 | 1 - 2 | 388 | 99 | ||
| XM_005248715.4 | XP_005248772.1 | 1 - 2 | 388 | 99 | ||
| GenBank mRNA | AK097021.1 | - | 1 - 2 | 257 | 99 | |
| AK290232.1 | - | 1 - 2 | 129 | 99 | ||
| AL832009.2 | - | 1 - 2 | 217 | 99 | ||
| BC136443.1 | - | 1 - 2 | 172 | 9020432 | 99 | |
| BX647178.1 | - | 1 - 2 | 368 | 99 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001882.hg
Additional Information:
This assay is in the 5'UTR region of the AK097021.1,AK290232.1,AL832009.2,BC136443.1,NM_001300866.1,NM_015687.3,XM_005248713.3,XM_005248715.4 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000498523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000498523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000498523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000498523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000498523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | ENST00000498523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NR_110608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NR_110608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NR_110608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600014316.hg.1 | NR_110608.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022958.hg.1 | TC06001882.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022958.hg.1 | ENST00000393004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022958.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022957.hg.1 | TC06001882.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022957.hg.1 | ENST00000237172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022957.hg.1 | ENST00000393004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022957.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022954.hg.1 | TC06001882.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022954.hg.1 | ENST00000237172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022954.hg.1 | ENST00000393004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022954.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022956.hg.1 | TC06001882.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022956.hg.1 | ENST00000237172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022956.hg.1 | ENST00000393004.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06022956.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202191.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202191.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202191.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202191.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202191.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600202191.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202191.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202191.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600202191.hg.1 | TC0600014316.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000237172.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000370020.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000498523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000498523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000498523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000498523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000498523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | ENST00000498523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001289987.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_001300866.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NM_015687.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NR_110608.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NR_110608.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NR_110608.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600014316.hg.1 | NR_110608.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 113585 | - | - |

FILIP1
Genomic Map