| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024244.hg.1 |
TC07001601.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024244.hg.1 |
ENST00000248633.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024244.hg.1 |
ENST00000428214.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024244.hg.1 |
ENST00000438045.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024244.hg.1 |
ENST00000541751.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024244.hg.1 |
ENST00000545192.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024244.hg.1 |
NM_000466.2 |
Refseq |