Assay Details
| Gene Symbol: SLC7A10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019849.2 | NP_062823.1 | 2 - 3 | 502 | 115 | |
| XM_006723284.2 | XP_006723347.1 | 2 - 3 | 374 | 115 | ||
| XM_011527119.1 | XP_011525421.1 | 2 - 3 | 374 | 115 | ||
| GenBank mRNA | AB037670.1 | - | 2 - 3 | 454 | 115 | |
| AJ277731.1 | - | 2 - 3 | 463 | 115 | ||
| AK316594.1 | - | 2 - 3 | 374 | 115 | ||
| BC035627.1 | - | 2 - 3 | 445 | 5200601 | 115 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001395.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
amino acid transmembrane transport
amino acid transport
neutral amino acid transport
L-serine transport
D-alanine transport
D-serine transport
leukocyte migration
neutral amino acid transmembrane transporter activity
L-serine transmembrane transporter activity
antiporter activity
amino acid transmembrane transport
amino acid transport
neutral amino acid transport
L-serine transport
D-alanine transport
D-serine transport
leukocyte migration
neutral amino acid transmembrane transporter activity
L-serine transmembrane transporter activity
antiporter activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19019247.hg.1 | ENST00000253188.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19019247.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19019247.hg.1 | TC19001395.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000587064.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000587064.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000587064.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000587064.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000587064.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000587064.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000587064.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000587064.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000587064.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000587064.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000587064.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000587064.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000590490.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900010379.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900185078.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185078.hg.1 | ENST00000587064.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185078.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185078.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185078.hg.1 | NM_019849.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900185078.hg.1 | TC1900010379.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900185078.hg.1 | ENST00000253188.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185078.hg.1 | ENST00000587064.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185078.hg.1 | ENST00000590036.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185078.hg.1 | ENST00000592596.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900185078.hg.1 | NM_019849.2 | Refseq |

SLC7A10
Genomic Map