| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000555170.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000555170.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000555170.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000555933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000555933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000555933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000555933.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000557456.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000557456.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000557456.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000557456.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
ENST00000557456.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14012838.hg.1 |
ENST00000298355.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14012838.hg.1 |
ENST00000338969.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14012838.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14012838.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14012838.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR14012838.hg.1 |
TC14001127.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000555170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000555170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000555170.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000555933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000555933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000555933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000555933.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000557456.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000557456.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000557456.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000557456.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
ENST00000557456.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1400009151.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400167611.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400167611.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400167611.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400167611.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400167611.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400167611.hg.1 |
ENST00000298355.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400167611.hg.1 |
ENST00000338969.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400167611.hg.1 |
ENST00000360392.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1400167611.hg.1 |
NM_015163.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400167611.hg.1 |
NM_052978.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1400167611.hg.1 |
TC1400009151.hg.1 |
Clariom D TC |