Assay Details
| Gene Symbol: TRIM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015163.5 | NP_055978.4 | 8 - 9 | 2678 | 69 | |
| XM_006720018.3 | XP_006720081.1 | 12 - 13 | 3538 | 69 | ||
| XM_011536389.2 | XP_011534691.1 | 12 - 13 | 3541 | 69 | ||
| XM_017020943.1 | XP_016876432.1 | 12 - 13 | 3526 | 69 | ||
| XM_017020944.1 | XP_016876433.1 | 11 - 12 | 3349 | 69 | ||
| XM_017020945.1 | XP_016876434.1 | 9 - 10 | 3283 | 69 | ||
| XM_017020946.1 | XP_016876435.1 | 9 - 10 | 3271 | 69 | ||
| XM_017020949.1 | XP_016876438.1 | 7 - 8 | 2986 | 69 | ||
| GenBank mRNA | AK055388.1 | - | 11 - 12 | 2546 | 69 | |
| BC063872.1 | - | 8 - 9 | 3035 | 6471552 | 69 | |
| D87458.2 | - | 8 - 9 | 2054 | 69 | ||
| DA173440.1 | - | 1 - 2 | 122 | 69 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001127.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
synaptic vesicle exocytosis
protein ubiquitination
negative regulation of SNARE complex assembly
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of calcium ion-dependent exocytosis
SNARE binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
protein homodimerization activity
synaptic vesicle exocytosis
protein ubiquitination
negative regulation of SNARE complex assembly
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of calcium ion-dependent exocytosis
SNARE binding
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity
protein homodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000555170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000555170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000555170.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012824.hg.1 | ENST00000298355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012824.hg.1 | ENST00000338969.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012824.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012824.hg.1 | TC14001127.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400167595.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167595.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167595.hg.1 | ENST00000555170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167595.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167595.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167595.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167595.hg.1 | ENST00000555170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167595.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167595.hg.1 | TC1400009151.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400167594.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167594.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167594.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167594.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167594.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167594.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167594.hg.1 | TC1400009151.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000298355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000338969.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000360392.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000555170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000555170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000555170.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000555933.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | ENST00000557456.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_015163.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009151.hg.1 | NM_052978.4 | Refseq |

TRIM9
Genomic Map