Assay Details
| Gene Symbol: FAM160A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098794.1 | NP_001092264.1 | 3 - 4 | 1136 | 79 | |
| NM_032127.3 | NP_115503.2 | 3 - 4 | 1136 | 79 | ||
| XM_006718343.2 | XP_006718406.1 | 3 - 4 | 1058 | 79 | ||
| XM_011520397.2 | XP_011518699.1 | 3 - 4 | 1072 | 79 | ||
| XM_011520398.2 | XP_011518700.1 | 3 - 4 | 1117 | 79 | ||
| XM_011520399.2 | XP_011518701.1 | 3 - 4 | 1117 | 79 | ||
| GenBank mRNA | AB051546.1 | - | 3 - 4 | 1041 | 79 | |
| AK056468.1 | - | 3 - 4 | 1088 | 79 | ||
| AK291702.1 | - | 3 - 4 | 1061 | 79 | ||
| AL136723.1 | - | 3 - 4 | 1070 | 79 | ||
| BC030825.1 | - | 3 - 4 | 1049 | 4811942 | 79 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001353.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11016950.hg.1 | ENST00000265978.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11016950.hg.1 | ENST00000442917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11016950.hg.1 | ENST00000449352.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11016950.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11016950.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11016950.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11016950.hg.1 | TC11001353.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000532797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | ENST00000532797.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000529360.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000532797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | ENST00000532797.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009962.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175108.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175108.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175108.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175108.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175108.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175108.hg.1 | ENST00000265978.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175108.hg.1 | ENST00000449352.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175108.hg.1 | ENST00000524416.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100175108.hg.1 | NM_001098794.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175108.hg.1 | NM_032127.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100175108.hg.1 | TC1100009962.hg.1 | Clariom D TC |

FAM160A2
Genomic Map