Assay Details
| Gene Symbol: GRIK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000831.3 | NP_000822.2 | 5 - 6 | 918 | 71 | |
| GenBank mRNA | AJ249210.1 | - | 5 - 6 | 804 | 71 | |
| AJ299451.1 | - | 5 - 6 | 783 | 71 | ||
| U16127.1 | - | 5 - 6 | 801 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002513.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
adenylate cyclase-inhibiting G-protein coupled glutamate receptor signaling pathway
glutamate receptor signaling pathway
G-protein coupled glutamate receptor signaling pathway
ion transmembrane transport
ionotropic glutamate receptor signaling pathway
regulation of membrane potential
negative regulation of synaptic transmission, glutamatergic
adenylate cyclase inhibiting G-protein coupled glutamate receptor activity
G-protein coupled receptor binding
ionotropic glutamate receptor activity
extracellular-glutamate-gated ion channel activity
glutamate receptor activity
ligand-gated ion channel activity
kainate selective glutamate receptor activity
adenylate cyclase-inhibiting G-protein coupled glutamate receptor signaling pathway
glutamate receptor signaling pathway
G-protein coupled glutamate receptor signaling pathway
ion transmembrane transport
ionotropic glutamate receptor signaling pathway
regulation of membrane potential
negative regulation of synaptic transmission, glutamatergic
adenylate cyclase inhibiting G-protein coupled glutamate receptor activity
G-protein coupled receptor binding
ionotropic glutamate receptor activity
extracellular-glutamate-gated ion channel activity
glutamate receptor activity
ligand-gated ion channel activity
kainate selective glutamate receptor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039054.hg.1 | TC01002513.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039054.hg.1 | ENST00000373091.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039054.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01039054.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202412.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202412.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202412.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202412.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202412.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100202412.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100202412.hg.1 | TC0100013730.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373091.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000373093.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000462621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | ENST00000479620.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013730.hg.1 | NM_000831.3 | Refseq |

GRIK3
Genomic Map