Assay Details
| Gene Symbol: KANK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198471.2 | NP_940873.2 | 3 - 4 | 1395 | 130 | |
| XM_006722718.3 | XP_006722781.1 | 3 - 4 | 1514 | 130 | ||
| XM_011527884.2 | XP_011526186.1 | 3 - 4 | 1771 | 130 | ||
| XM_017026563.1 | XP_016882052.1 | 2 - 3 | 1531 | 130 | ||
| GenBank mRNA | AB284126.1 | - | 3 - 4 | 1395 | 130 | |
| AK128815.1 | - | 3 - 4 | 1395 | 130 | ||
| BC066124.1 | - | 3 - 4 | 1395 | 30345663 | 130 | |
| BC066775.1 | - | 3 - 4 | 1395 | 30333203 | 130 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001129.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015546.hg.1 | ENST00000330915.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015546.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015546.hg.1 | TC19001129.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015547.hg.1 | ENST00000330915.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015547.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015547.hg.1 | TC19001129.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015548.hg.1 | ENST00000330915.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015548.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015548.hg.1 | TC19001129.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015544.hg.1 | ENST00000330915.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015544.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19015544.hg.1 | TC19001129.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900009528.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176654.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176654.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176654.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176654.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176654.hg.1 | TC1900009528.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900176654.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176654.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176654.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176654.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176652.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176652.hg.1 | ENST00000593331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176652.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176652.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176652.hg.1 | TC1900009528.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900176652.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176652.hg.1 | ENST00000593331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176652.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176652.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176655.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176655.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176655.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176655.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176655.hg.1 | TC1900009528.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900176655.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176655.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176655.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176655.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176653.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176653.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176653.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176653.hg.1 | TC1900009528.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900176653.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176653.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176653.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176656.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176656.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176656.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176656.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176656.hg.1 | TC1900009528.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900176656.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176656.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176656.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176656.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176650.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176650.hg.1 | ENST00000593331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176650.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176650.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176650.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176650.hg.1 | NM_198471.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900176650.hg.1 | TC1900009528.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900176650.hg.1 | ENST00000330915.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176650.hg.1 | ENST00000593331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176650.hg.1 | ENST00000593649.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176650.hg.1 | ENST00000595639.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176650.hg.1 | ENST00000610351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900176650.hg.1 | NM_198471.2 | Refseq |

KANK3
Genomic Map