Assay Details
| Gene Symbol: MARK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286124.1 | NP_001273053.1 | 3 - 4 | 940 | 178 | |
| NM_001286126.1 | NP_001273055.1 | 3 - 4 | 940 | 178 | ||
| NM_018650.4 | NP_061120.3 | 3 - 4 | 940 | 178 | ||
| XM_005273134.4 | XP_005273191.1 | 3 - 4 | 807 | 178 | ||
| XM_011509561.2 | XP_011507863.1 | 3 - 4 | 936 | 178 | ||
| XM_017001305.1 | XP_016856794.1 | 4 - 5 | 397 | 178 | ||
| XM_017001307.1 | XP_016856796.1 | 4 - 5 | 837 | 178 | ||
| GenBank mRNA | AF154845.1 | - | 3 - 4 | 566 | 178 | |
| AK295506.1 | - | 3 - 4 | 438 | 178 | ||
| AK314614.1 | - | 3 - 4 | 600 | 178 | ||
| BC113869.1 | - | 3 - 4 | 476 | 40036731 | 178 | |
| BC114478.1 | - | 3 - 4 | 476 | 40036730 | 178 | |
| KJ534879.1 | - | 3 - 4 | 300 | 178 | ||
| KJ534881.1 | - | 3 - 4 | 300 | 178 | ||
| KJ535089.1 | - | 3 - 4 | 299 | 178 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017001305.1, XM_017001307.1 transcripts.
Set Membership: |
|

MARK1
Genomic Map