| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03028635.hg.1 |
TC03001583.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03028635.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03028635.hg.1 |
ENST00000398399.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03028635.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03028636.hg.1 |
TC03001583.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03028636.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03028636.hg.1 |
ENST00000398399.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03028636.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000494229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000494229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
ENST00000494229.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300187151.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187151.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187151.hg.1 |
TC0300011706.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0300187154.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187154.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187154.hg.1 |
ENST00000494229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187154.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300187154.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187154.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187154.hg.1 |
ENST00000494229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187154.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300187154.hg.1 |
TC0300011706.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000494229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000494229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
ENST00000494229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0300011706.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300187153.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187153.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187153.hg.1 |
ENST00000494229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187153.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300187153.hg.1 |
ENST00000383698.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187153.hg.1 |
ENST00000398399.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187153.hg.1 |
ENST00000494229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300187153.hg.1 |
NM_016206.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300187153.hg.1 |
TC0300011706.hg.1 |
Clariom D TC |