Assay Details
| Gene Symbol: LOC102724053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125873.1 | 1 - 2 | 245 | 149 | ||
| GenBank mRNA | AA437152.1 | - | 1 - 2 | 244 | 148 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724053| Gene Symbol: LOC102724053 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125873.1 | 1 - 2 | 245 | 149 | ||
| GenBank mRNA | AA437152.1 | - | 1 - 2 | 244 | 148 | |
Set Membership: |
|